Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TAPT1-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027696.1 | 3 | 755 | |||
NR_027697.1 | 3 | 704 | ||||
GenBank mRNA | BF732723.1 | - | 3 | 378 | ||
DA640966.1 | - | 3 | 378 | |||
DA797317.1 | - | 1 | 16 |