Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other TTC28-AS1 SIRNA Assays ›
          Gene Symbol:
          TTC28-AS1
          Gene Name:
          TTC28 antisense RNA 1
          Gene Aliases:
          TTC28-AS, TTC28AS, dJ353E16.2
          Chromosome Location:
          Chr.22: 27919376 - 28002679 on Build GRCh38
          UniGene ID:
          Hs.602319
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID n542296

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: TTC28-AS1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_026962.1 1, 2 168
          GenBank mRNA BC016779.1 - 1, 2 78
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA BF243724.1 - 75 Chr. - - [-] on Build
          GenBankmRNA DA779688.1 - 1, 2 87 Chr. 22 - 27919457 - 27922518 [+] on Build GRCh38
          GenBankmRNA HY096240.1 - 1, 2 77 Chr. 22 - 27919468 - 27922404 [+] on Build GRCh38
          GenBankmRNA BU858805.1 - 1, 2 105 Chr. 22 - 27919442 - 27922154 [+] on Build GRCh38
          GenBankmRNA DA779683.1 - 1, 2 89 Chr. 22 - 27919455 - 27922516 [+] on Build GRCh38
          GenBankmRNA BF773185.1 - 211 Chr. - - [-] on Build
          GenBankmRNA DA503708.1 - 1, 2 78 Chr. 22 - 27919466 - 27922485 [+] on Build GRCh38
          GenBankmRNA DA121660.1 - 1, 2 87 Chr. 22 - 27919457 - 27922461 [+] on Build GRCh38
          GenBankmRNA DB444867.1 - 1, 2 63 Chr. 22 - 27919481 - 27922380 [+] on Build GRCh38
          GenBankmRNA DW434093.1 - 1, 2 89 Chr. 22 - 27919467 - 27922178 [+] on Build GRCh38
          GenBankmRNA DB187377.1 - 1, 2 87 Chr. 22 - 27919457 - 27922484 [+] on Build GRCh38
          GenBankmRNA HY066944.1 - 1, 2 77 Chr. 22 - 27919468 - 27922395 [+] on Build GRCh38
          GenBankmRNA HY172362.1 - 1, 2 111 Chr. 22 - 27919437 - 27922366 [+] on Build GRCh38
          GenBankmRNA DA124925.1 - 1, 2 78 Chr. 22 - 27919466 - 27922496 [+] on Build GRCh38
          GenBankmRNA DA870155.1 - 1, 2 107 Chr. 22 - 27919437 - 27922446 [+] on Build GRCh38
          GenBankmRNA DB074404.1 - 1, 2 74 Chr. 22 - 27919470 - 27922485 [+] on Build GRCh38
          GenBankmRNA HY077858.1 - 1, 2 77 Chr. 22 - 27919468 - 27922411 [+] on Build GRCh38
          GenBankmRNA HY029971.1 - 1, 2 108 Chr. 22 - 27919437 - 27924525 [+] on Build GRCh38
          GenBankmRNA DA126837.1 - 1, 2 74 Chr. 22 - 27919470 - 27922519 [+] on Build GRCh38
          GenBankmRNA DB045006.1 - 1, 2 52 Chr. 22 - 27919493 - 27922531 [+] on Build GRCh38
          GenBankmRNA HY084175.1 - 1, 2 77 Chr. 22 - 27919468 - 27922414 [+] on Build GRCh38

          Back To Top

          More Information



          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0