Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other SNHG20 SIRNA Assays ›
          Gene Symbol:
          SNHG20
          Gene Name:
          small nucleolar RNA host gene 20
          Gene Aliases:
          C17orf86, LINC00338, NCRNA00338, SCARNA16HG
          Chromosome Location:
          Chr.17: 77088643 - 77094986 on Build GRCh38
          UniGene ID:
          Hs.720923
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID n543198

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: SNHG20
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_027058.1 3 528
          GenBank mRNA AF130050.1 - 2 153
          DA068554.1 - 3 416
          DA916381.1 - 3 528
          FN357115.1 - 3 369
          FN357116.1 - 3 239
          FN357117.1 - 3 422
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA DA012348.1 - 3 403 Chr. 17 - 77088765 - 77093499 [+] on Build GRCh38
          GenBankmRNA DA982200.1 - 3 428 Chr. 17 - 77088733 - 77093481 [+] on Build GRCh38
          GenBankmRNA DA436888.1 - 3 511 Chr. 17 - 77088660 - 77093418 [+] on Build GRCh38
          GenBankmRNA DA156398.1 - 3 407 Chr. 17 - 77088755 - 77093501 [+] on Build GRCh38
          GenBankmRNA DA067075.1 - 3 416 Chr. 17 - 77088755 - 77093501 [+] on Build GRCh38
          GenBankmRNA DA911256.1 - 3 509 Chr. 17 - 77088662 - 77093395 [+] on Build GRCh38
          GenBankmRNA DA513395.1 - 3 457 Chr. 17 - 77088719 - 77093444 [+] on Build GRCh38
          GenBankmRNA DA887580.1 - 3 416 Chr. 17 - 77088755 - 77093490 [+] on Build GRCh38
          GenBankmRNA DA075642.1 - 3 476 Chr. 17 - 77088695 - 77093445 [+] on Build GRCh38
          GenBankmRNA DB028110.1 - 3 417 Chr. 17 - 77088755 - 77093459 [+] on Build GRCh38
          GenBankmRNA DA684608.1 - 3 352 Chr. 17 - 77088819 - 77093530 [+] on Build GRCh38
          GenBankmRNA DA609465.1 - 3 412 Chr. 17 - 77088762 - 77093485 [+] on Build GRCh38
          GenBankmRNA DB175957.1 - 3 478 Chr. 17 - 77088693 - 77093438 [+] on Build GRCh38
          GenBankmRNA AI065041.1 - 2 153 Chr. 17 - 77089221 - 77093758 [+] on Build GRCh38
          GenBankmRNA DA024063.1 - 3 331 Chr. 17 - 77088840 - 77093583 [+] on Build GRCh38
          GenBankmRNA DA840981.1 - 3 486 Chr. 17 - 77088685 - 77093419 [+] on Build GRCh38
          GenBankmRNA DA926940.1 - 3 480 Chr. 17 - 77088695 - 77093446 [+] on Build GRCh38
          GenBankmRNA DA084161.1 - 3 517 Chr. 17 - 77088654 - 77093386 [+] on Build GRCh38
          GenBankmRNA AL530240.3 - 3 447 Chr. 17 - 77088723 - 77093777 [+] on Build GRCh38
          GenBankmRNA DA163899.1 - 3 511 Chr. 17 - 77088660 - 77093404 [+] on Build GRCh38
          GenBankmRNA DA283845.1 - 3 512 Chr. 17 - 77088660 - 77093421 [+] on Build GRCh38
          GenBankmRNA DB171108.1 - 3 416 Chr. 17 - 77088755 - 77093523 [+] on Build GRCh38
          GenBankmRNA DA818222.1 - 3 476 Chr. 17 - 77088695 - 77093426 [+] on Build GRCh38
          GenBankmRNA CX788271.1 - 3 463 Chr. 17 - 77088708 - 77093561 [+] on Build GRCh38

          Back To Top

          More Information



          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-865d7754c7-mcq8j:80/100.66.130.62:80.
          git-commit: 7ff36f61b459eb8f02ba5a263f0e3ce367ca82d3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0