Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC102724663 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_120491.1 | 2 | 176 | |||
GenBank mRNA | BG186381.1 | - | 2 | 180 |