Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other MIR9-3HG SIRNA Assays ›
          Gene Symbol:
          MIR9-3HG
          Gene Name:
          MIR9-3 host gene
          Gene Aliases:
          LINC00925
          Chromosome Location:
          Chr.15: 89361579 - 89398487 on Build GRCh38
          UniGene ID:
          Hs.136313
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID n544672

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: MIR9-3HG
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_015411.1 5 545
          NR_133001.1 5 1125
          NR_133002.1 4 426
          NR_133003.1 5 460
          GenBank mRNA AK054931.1 - 5 460
          AK123792.1 - 2 1312
          AK125292.1 - 5 471
          BX441353.2 - 4 458
          DA059714.1 - 5 460
          DB020558.1 - 3 297
          DB067527.1 - 3 235
          DB068800.1 - 3 235
          DB452761.1 - 3 260
          DB454083.1 - 3 235
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA BI825996.1 - 3 231 Chr. 15 - 89379130 - 89395344 [+] on Build GRCh38
          GenBankmRNA DB097661.1 - 3 238 Chr. 15 - 89379123 - 89395193 [+] on Build GRCh38
          GenBankmRNA DB027914.1 - 3 236 Chr. 15 - 89379126 - 89395172 [+] on Build GRCh38
          GenBankmRNA DB075548.1 - 4 285 Chr. 15 - 89379124 - 89395119 [+] on Build GRCh38
          GenBankmRNA HY110386.1 - 4 394 Chr. 15 - 89371610 - 89389423 [+] on Build GRCh38
          GenBankmRNA HY004906.1 - 3 280 Chr. 15 - 89379081 - 89389475 [+] on Build GRCh38
          GenBankmRNA DB061891.1 - 3 239 Chr. 15 - 89379123 - 89395144 [+] on Build GRCh38
          GenBankmRNA DB036793.1 - 3 322 Chr. 15 - 89379038 - 89395091 [+] on Build GRCh38
          GenBankmRNA DB071179.1 - 3 323 Chr. 15 - 89379039 - 89395043 [+] on Build GRCh38
          GenBankmRNA DB021481.1 - 3 323 Chr. 15 - 89379039 - 89395082 [+] on Build GRCh38
          GenBankmRNA DB068534.1 - 3 149 Chr. 15 - 89378106 - 89395219 [+] on Build GRCh38
          GenBankmRNA HY056341.1 - 3 307 Chr. 15 - 89379054 - 89389426 [+] on Build GRCh38
          GenBankmRNA DB066190.1 - 3 144 Chr. 15 - 89378110 - 89395222 [+] on Build GRCh38
          GenBankmRNA BG819916.1 - 2 88 Chr. 15 - 89387817 - 89395478 [+] on Build GRCh38
          GenBankmRNA DA451099.1 - 3 151 Chr. 15 - 89378106 - 89395242 [+] on Build GRCh38
          GenBankmRNA DB060384.1 - 3 238 Chr. 15 - 89379123 - 89395137 [+] on Build GRCh38
          GenBankmRNA DB068926.1 - 3 254 Chr. 15 - 89379107 - 89395137 [+] on Build GRCh38
          GenBankmRNA DB035646.1 - 3 254 Chr. 15 - 89379107 - 89395151 [+] on Build GRCh38
          GenBankmRNA DB058234.1 - 3 238 Chr. 15 - 89379123 - 89395134 [+] on Build GRCh38
          GenBankmRNA DA804092.1 - 3 155 Chr. 15 - 89378102 - 89395217 [+] on Build GRCh38
          GenBankmRNA DB035856.1 - 3 323 Chr. 15 - 89379038 - 89395086 [+] on Build GRCh38
          GenBankmRNA DA340519.1 - 3 242 Chr. 15 - 89379119 - 89395185 [+] on Build GRCh38
          GenBankmRNA DB021867.1 - 3 239 Chr. 15 - 89379124 - 89395144 [+] on Build GRCh38
          GenBankmRNA DB042606.1 - 3 242 Chr. 15 - 89379119 - 89395151 [+] on Build GRCh38
          GenBankmRNA DB023242.1 - 3 126 Chr. 15 - 89378131 - 89395268 [+] on Build GRCh38

          Back To Top

          More Information



          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6659c69d8f-875ns:80/100.66.131.9:80.
          git-commit: 1827ea18100b76eacea45a9906e00cdefce2d263
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.30.1-2025.05.22-1.0