Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC283038 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033848.1 | 5 | 1901 | |||
GenBank mRNA | BC033536.1 | - | 5 | 1900 | ||
BC048981.1 | - | 5 | 1899 |