Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ENTPD1-AS1 SIRNA Assays ›
          Gene Symbol:
          ENTPD1-AS1
          Gene Name:
          ENTPD1 antisense RNA 1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.10: 95753206 - 96090235 on Build GRCh38
          UniGene ID:
          Hs.538374
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID n548179

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: ENTPD1-AS1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_038444.1 1 223
          NR_134320.1 1 223
          NR_134321.1 1 223
          NR_134322.1 1 223
          GenBank mRNA AK091396.1 - 1 202
          AK096469.1 - 1 200
          AL120918.1 - 1 161
          BC028619.1 - 1 222
          BU852745.1 - 1 226
          DA419635.1 - 1 203
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA BU854016.1 - 228 Chr. - - [-] on Build
          GenBankmRNA DB445857.1 - 1 209 Chr. 10 - 96057830 - 96090220 [-] on Build GRCh38
          GenBankmRNA BP871689.1 - 1 203 Chr. 10 - 96042608 - 96090215 [-] on Build GRCh38
          GenBankmRNA AV762020.1 - 1 200 Chr. 10 - 96055291 - 96090212 [-] on Build GRCh38
          GenBankmRNA BI225716.1 - 1 142 Chr. 10 - 95861313 - 96090155 [-] on Build GRCh38
          GenBankmRNA BI226415.1 - 1 142 Chr. 10 - 95861295 - 96090155 [-] on Build GRCh38
          GenBankmRNA BM549802.1 - 1 206 Chr. 10 - 95875320 - 96090202 [-] on Build GRCh38
          GenBankmRNA DA213255.1 - 1 203 Chr. 10 - 95876619 - 96090214 [-] on Build GRCh38
          GenBankmRNA BE513929.1 - 1 123 Chr. 10 - 95861550 - 96090135 [-] on Build GRCh38
          GenBankmRNA AW732710.1 - 1 127 Chr. 10 - 95876567 - 96090139 [-] on Build GRCh38
          GenBankmRNA BP871686.1 - 1 203 Chr. 10 - 96042601 - 96090215 [-] on Build GRCh38
          GenBankmRNA BE396593.1 - 1 123 Chr. 10 - 95861500 - 96090135 [-] on Build GRCh38
          GenBankmRNA CR739709.1 - 1 186 Chr. 10 - 96021352 - 96090196 [-] on Build GRCh38
          GenBankmRNA CD707141.1 - 1 245 Chr. 10 - 96089207 - 96090241 [-] on Build GRCh38
          GenBankmRNA DA491898.1 - 1 202 Chr. 10 - 95785184 - 96090214 [-] on Build GRCh38
          GenBankmRNA DA769118.1 - 1 200 Chr. 10 - 95921342 - 96090212 [-] on Build GRCh38
          GenBankmRNA BE396777.1 - 1 122 Chr. 10 - 95861329 - 96090134 [-] on Build GRCh38

          Back To Top

          More Information



          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0