Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC171391 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_126342.1 | 4 | 456 | |||
NR_126343.1 | 2 | 702 | ||||
GenBank mRNA | BF529437.1 | - | 1 | 475 | ||
BU156985.1 | - | 4 | 426 |