Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MIR34AHG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_132742.1 | 2 | 703 | |||
GenBank mRNA | EF570048.1 | - | 2 | 470 | ||
EF592573.1 | - | 2 | 758 |