Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other SNHG21 SIRNA Assays ›
          Gene Symbol:
          SNHG21
          Gene Name:
          small nucleolar RNA host gene 21
          Gene Aliases:
          FSD2-AS1, SCARNA15HG
          Chromosome Location:
          Chr.15: 82750564 - 82757206 on Build GRCh38
          UniGene ID:
          Hs.513091
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID n550475

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: SNHG21
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_110096.1 1 225
          NR_110097.1 1 225
          NR_110098.1 1 225
          NR_110099.1 1 225
          GenBank mRNA BX112407.1 - 1 202
          CF129490.1 - 1 206
          CK904397.1 - 1 205
          DA396616.1 - 1 217
          DB154257.1 - 1 225
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA AA287242.1 - 1 97 Chr. 15 - 82750703 - 82757078 [+] on Build GRCh38
          GenBankmRNA N48229.1 - 1 224 Chr. 15 - 82750564 - 82756764 [+] on Build GRCh38
          GenBankmRNA BQ188589.1 - 1 42 Chr. 15 - 82750749 - 82757208 [+] on Build GRCh38
          GenBankmRNA BX482265.1 - 1 190 Chr. 15 - 82750599 - 82755058 [+] on Build GRCh38
          GenBankmRNA BI912731.1 - 195 Chr. - - [-] on Build
          GenBankmRNA BM054859.1 - 1 189 Chr. 15 - 82750600 - 82757096 [+] on Build GRCh38
          GenBankmRNA DA907531.1 - 1 218 Chr. 15 - 82750572 - 82754911 [+] on Build GRCh38
          GenBankmRNA DB148883.1 - 1 218 Chr. 15 - 82750572 - 82754869 [+] on Build GRCh38
          GenBankmRNA R07313.1 - 201 Chr. - - [-] on Build
          GenBankmRNA BU950025.1 - 1 153 Chr. 15 - 82750636 - 82756974 [+] on Build GRCh38
          GenBankmRNA BX538250.1 - 1 218 Chr. 15 - 82750572 - 82757085 [+] on Build GRCh38
          GenBankmRNA DA955724.1 - 1 225 Chr. 15 - 82750566 - 82754922 [+] on Build GRCh38
          GenBankmRNA HY070763.1 - 1 218 Chr. 15 - 82750574 - 82754751 [+] on Build GRCh38
          GenBankmRNA AL555315.3 - 1 185 Chr. 15 - 82750604 - 82754784 [+] on Build GRCh38
          GenBankmRNA DW429916.1 - 1 240 Chr. 15 - 82750562 - 82754722 [+] on Build GRCh38
          GenBankmRNA DA299174.1 - 1 223 Chr. 15 - 82750569 - 82754903 [+] on Build GRCh38
          GenBankmRNA DA079565.1 - 1 227 Chr. 15 - 82750564 - 82754918 [+] on Build GRCh38
          GenBankmRNA EL946018.1 - 1 59 Chr. 15 - 82750730 - 82757129 [+] on Build GRCh38
          GenBankmRNA DB185706.1 - 1 218 Chr. 15 - 82750573 - 82754926 [+] on Build GRCh38
          GenBankmRNA DB141074.1 - 1 206 Chr. 15 - 82750583 - 82754826 [+] on Build GRCh38
          GenBankmRNA BM829432.1 - 1 217 Chr. 15 - 82750572 - 82756992 [+] on Build GRCh38

          Back To Top

          More Information



          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0