Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ivns1abp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039511.1 | NP_001034600.1 | 5 | 801 | ||
NM_001039512.1 | NP_001034601.1 | 5 | 801 | |||
NM_054102.2 | NP_473443.2 | 5 | 801 | |||
GenBank mRNA | AB055737.1 | - | 5 | 797 | ||
AB055738.1 | - | 5 | 797 | |||
AK007291.1 | - | 5 | 751 | |||
AK129229.1 | - | 5 | 739 | |||
AK144619.1 | - | 5 | 780 | |||
AK144835.1 | - | 5 | 782 | |||
AK145071.1 | - | 5 | 792 | |||
AK145845.1 | - | 5 | 751 | |||
AK146001.1 | - | 5 | 782 | |||
AK146329.1 | - | 2 | 125 | |||
AK146494.1 | - | 5 | 754 | |||
AK146645.1 | - | 5 | 752 | |||
AK159254.1 | - | 5 | 783 | |||
AK179875.1 | - | 4 | 353 | |||
AK181493.1 | - | 4 | 378 | |||
BC004092.1 | - | 4 | 501 | 3593827 | ||
BC040250.1 | - | 5 | 793 | |||
DQ914522.1 | - | 5 | 753 | |||
DQ914523.1 | - | 5 | 723 |