Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other CDK16 SIRNA Assays ›
          Gene Symbol:
          CDK16
          Gene Name:
          cyclin dependent kinase 16
          Gene Aliases:
          PCTAIRE, PCTAIRE1, PCTGAIRE, PCTK1
          Chromosome Location:
          Chr.X: 47217881 - 47229997 on Build GRCh38
          UniGene ID:
          Hs.496068
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s10158
          Validated

          Purification
          Size
          Availability Inventoried
          Catalog # 4390824
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: CDK16
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001170460.1 NP_001163931.1 15 1690
          NM_006201.4 NP_006192.1 15 1825
          NM_033018.3 NP_148978.2 15 1932
          XM_011543920.2 XP_011542222.1 15 1702
          XM_011543921.1 XP_011542223.1 16 2097
          XM_011543922.1 XP_011542224.1 16 2085
          XM_011543923.1 XP_011542225.1 15 2015
          XM_011543924.1 XP_011542226.1 15 1825
          XM_011543925.2 XP_011542227.1 16 1907
          XM_011543926.1 XP_011542228.1 15 2868
          XM_011543927.2 XP_011542229.1 16 1945
          XM_011543928.1 XP_011542230.1 16 2300
          XM_017029569.1 XP_016885058.1 16 2097
          XM_017029570.1 XP_016885059.1 16 1907
          XM_017029571.1 XP_016885060.1 15 2868
          XM_017029572.1 XP_016885061.1 16 1945
          XM_017029573.1 XP_016885062.1 16 2300
          GenBank mRNA AB209465.1 - 12 2062
          AK096881.1 - 15 1815
          AK131486.1 - 11 2607
          AK301832.1 - 15 1690
          AK303849.1 - 16 1705
          BC001048.2 - 15 1828 3504276
          BC006190.2 - 15 1642 3830397
          BC009852.2 - 13 1088
          BC015607.2 - 15 1795 4762081
          BT006827.1 - 14 1390
          X66363.1 - 15 1513

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_031077, NM_001004132, XM_006256618, XM_006256619, XM_006256617, and others.
          This siRNA also targets the following transcripts in Mouse: NM_011049, X69025, AK143518, BC013663, AK141083, and others.

          Matching Taqman Gene Expression Assay:


          Hs00178837_m1

          Gene Ontology Categories:

          Function(s) Process(es)

          protein phosphorylation
          exocytosis
          spermatogenesis
          growth hormone secretion
          neuron projection development
          regulation of cell cycle
          regulation of insulin secretion involved in cellular response to glucose stimulus
          protein serine/threonine kinase activity
          cyclin-dependent protein serine/threonine kinase activity
          protein binding
          ATP binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0