Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 9530068E07Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153117.2 | NP_694757.1 | 3 | 763 | ||
GenBank mRNA | AK007314.1 | - | 3 | 625 | ||
AK079269.1 | - | 3 | 764 | |||
AK135934.1 | - | 3 | 727 | |||
AK144677.1 | - | 3 | 727 | |||
AK145254.1 | - | 3 | 732 | |||
AK147956.1 | - | 3 | 727 | |||
AK150517.1 | - | 3 | 747 | |||
AK154955.1 | - | 3 | 733 | |||
BC006957.1 | - | 3 | 617 | |||
BC034829.1 | - | 3 | 737 | 4482756 |