Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adgrg6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002268.3 | NP_001002268.1 | 5, 6 | 1269 | ||
GenBank mRNA | AB183545.1 | - | 5, 6 | 1270 |
Set Membership: |
![]() |