Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam222b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145430.2 | NP_663405.1 | 3 | 334 | ||
GenBank mRNA | AK085728.2 | - | 3 | 341 | ||
AK141119.1 | - | 3 | 296 | |||
BC017647.1 | - | 3 | 438 | 4527119 | ||
BC063099.1 | - | 2 | 230 | 6393931 |