Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pomt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153415.3 | NP_700464.2 | 13 | 1910 | ||
GenBank mRNA | AF246235.1 | - | 13 | 1514 | ||
AK043573.1 | - | 13 | 1611 | |||
AK043713.1 | - | 13 | 1452 | |||
AK050915.1 | - | 13 | 1479 | |||
AK076813.1 | - | 13 | 1912 | |||
AK214879.1 | - | 1 | 31 | |||
AY090481.1 | - | 13 | 1910 | |||
AY090482.1 | - | 13 | 1510 | |||
BC052045.1 | - | 13 | 1482 | 6402328 | ||
BC062965.1 | - | 13 | 1482 | |||
BC079589.1 | - | 13 | 1480 |