Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gmds | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146041.2 | NP_666153.1 | 7 | 743 | ||
GenBank mRNA | AK086076.1 | - | 7 | 787 | ||
BC031788.1 | - | 7 | 743 | 4165509 | ||
BC093502.1 | - | 7 | 767 | 4215865 |