Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spryd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033277.3 | NP_001028449.2 | 3 | 290 | ||
GenBank mRNA | AK146689.1 | - | 3 | 293 | ||
BC048820.1 | - | 3 | 267 | |||
BU057890.1 | - | 3 | 225 | |||
BY739425.1 | - | 3 | 290 |