Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc37a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242427.1 | NP_001229356.1 | 3 | 968 | ||
NM_153062.2 | NP_694702.1 | 3 | 548 | |||
GenBank mRNA | AK033656.1 | - | 3 | 437 | ||
AK143636.1 | - | 3 | 974 | |||
AK153706.1 | - | 3 | 971 | |||
AK163990.1 | - | 2 | 150 | |||
BC027294.1 | - | 3 | 548 | 3985469 |