Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyb561a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282064.1 | NP_001268993.1 | 4 | 1002 | ||
NM_001282065.1 | NP_001268994.1 | 3 | 1099 | |||
NM_001282067.1 | NP_001268996.1 | 5 | 1022 | |||
NM_201351.2 | NP_958739.1 | 4 | 1192 | |||
GenBank mRNA | AK040692.1 | - | 896 | |||
AK139533.1 | - | 4 | 1180 | |||
AK160179.1 | - | 3 | 803 | |||
AK168841.1 | - | 4 | 896 | |||
AK169729.1 | - | 4 | 898 | |||
AK170911.1 | - | 5 | 690 | |||
AK172638.1 | - | 4 | 709 | |||
AK215320.1 | - | 4 | 288 | |||
BC065078.1 | - | 4 | 626 | 6817093 |