Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other PIK3CG SIRNA Assays ›
          Gene Symbol:
          PIK3CG
          Gene Name:
          phosphatidylinositol-4,5-bisphosphate 3-kinase catalytic subunit gamma
          Gene Aliases:
          PI3CG, PI3K, PI3Kgamma, PIK3, p110gamma, p120-PI3K
          Chromosome Location:
          Chr.7: 106865278 - 106908978 on Build GRCh38
          UniGene ID:
          Hs.32942
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s10532

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: PIK3CG
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001282426.1 NP_001269355.1 5 2514
          NM_001282427.1 NP_001269356.1 5 2438
          NM_002649.3 NP_002640.2 5 2663
          XM_005250443.3 XP_005250500.1 5 2531
          XM_011516316.1 XP_011514618.1 5 2496
          XM_011516317.1 XP_011514619.1 5 2496
          XM_017012328.1 XP_016867817.1 5 2496
          GenBank mRNA AF327656.1 - 4 2353
          AK292684.1 - 5 2438
          AK313753.1 - 5 2421
          AY496423.1 - 1 30
          BC035683.1 - 5 2406 5749986
          BX648341.1 - 5 2496
          X83368.1 - 5 2673

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          angiogenesis
          cytokine production
          adaptive immune response
          dendritic cell chemotaxis
          positive regulation of acute inflammatory response
          respiratory burst involved in defense response
          protein phosphorylation
          phosphatidylinositol biosynthetic process
          endocytosis
          inflammatory response
          G-protein coupled receptor signaling pathway
          positive regulation of cytosolic calcium ion concentration
          T cell chemotaxis
          negative regulation of triglyceride catabolic process
          phosphatidylinositol 3-kinase signaling
          platelet activation
          neutrophil chemotaxis
          secretory granule localization
          regulation of cell adhesion mediated by integrin
          natural killer cell chemotaxis
          phosphatidylinositol-3-phosphate biosynthetic process
          T cell proliferation
          T cell activation
          mast cell degranulation
          positive regulation of MAP kinase activity
          innate immune response
          phosphatidylinositol phosphorylation
          positive regulation of protein kinase B signaling
          negative regulation of cardiac muscle contraction
          platelet aggregation
          cellular response to cAMP
          neutrophil extravasation
          hepatocyte apoptotic process
          regulation of calcium ion transmembrane transport
          negative regulation of fibroblast apoptotic process
          protein kinase activity
          protein serine/threonine kinase activity
          protein binding
          ATP binding
          1-phosphatidylinositol-3-kinase activity
          phosphatidylinositol 3-kinase activity
          1-phosphatidylinositol-4-phosphate 3-kinase activity
          ephrin receptor binding
          phosphatidylinositol-4,5-bisphosphate 3-kinase activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7699c4d8ff-7jgns:80/100.66.130.62:80.
          git-commit: c11338b6b8d815146c5ef25758b63739976b60cb
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-Offline