Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arhgef4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183019.2 | NP_898840.2 | 8 | 1337 | ||
GenBank mRNA | AK034013.1 | - | 8 | 1301 | ||
AK122448.2 | - | 4 | 392 | |||
AK134629.1 | - | 1300 | ||||
AK138598.1 | - | 8 | 1297 | |||
AK163424.1 | - | 8 | 1319 | |||
AK169957.1 | - | 8 | 1357 | |||
BC055804.1 | - | 8 | 1298 | |||
BC094608.1 | - | 4857 | ||||
BC139371.1 | - | 8 | 1320 | 8862393 | ||
BC139397.1 | - | 8 | 1320 | 8862419 |