Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arhgef4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183019.2 | NP_898840.2 | 4 | 707 | ||
GenBank mRNA | AK031310.1 | - | 4 | 690 | ||
AK034013.1 | - | 4 | 671 | |||
AK134629.1 | - | 670 | ||||
AK163424.1 | - | 4 | 689 | |||
AK169449.1 | - | 4 | 690 | |||
AK169957.1 | - | 4 | 727 | |||
BB609907.1 | - | 4 | 709 | |||
BC055804.1 | - | 4 | 669 | |||
BC094608.1 | - | 4227 | ||||
BC139371.1 | - | 4 | 690 | 8862393 | ||
BC139397.1 | - | 4 | 690 | 8862419 |