Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccnyl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001097644.1 | NP_001091113.1 | 4 | 201 | ||
GenBank mRNA | AK132782.1 | - | 556 |