Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 2310035C23Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029349.1 | NP_083625.1 | 5 | 1055 | ||
NM_173187.3 | NP_775279.2 | 6 | 1127 | |||
GenBank mRNA | AK035724.1 | - | 6 | 1129 | ||
AK040168.1 | - | 7 | 763 | |||
AK129368.1 | - | 5 | 1028 | |||
AK148005.1 | - | 6 | 1107 | |||
BC060194.1 | - | 5 | 300 | |||
BC117949.1 | - | 6 | 1061 | 40098078 | ||
BC117950.1 | - | 5 | 993 | 40098082 |