Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc25a51 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009949.3 | NP_001009949.3 | 3 | 672 | ||
GenBank mRNA | AK051139.1 | - | 668 | |||
AK146788.1 | - | 3 | 634 | |||
AK150362.1 | - | 3 | 630 | |||
AK151688.1 | - | 3 | 630 | |||
AK152171.1 | - | 3 | 630 | |||
AK156168.1 | - | 3 | 630 | |||
AK156342.1 | - | 3 | 631 | |||
AK165253.1 | - | 3 | 668 | |||
AK198170.1 | - | 1 | 114 | |||
AK213361.1 | - | 1 | 63 | |||
BC089000.1 | - | 2 | 567 | 6418170 |