Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sh3tc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_194344.2 | NP_919325.2 | 10 | 1545 | ||
GenBank mRNA | AK028482.1 | - | 6 | 1641 | ||
AK131143.1 | - | 4 | 410 | |||
AK150064.1 | - | 10 | 1323 | |||
AK155330.1 | - | 10 | 1307 | |||
AK159089.1 | - | 10 | 1324 | |||
AK167283.1 | - | 10 | 1246 | |||
AK170383.1 | - | 10 | 1378 | |||
AK170396.1 | - | 10 | 1325 | |||
AK171209.1 | - | 10 | 1302 | |||
AK171242.1 | - | 10 | 1302 | |||
BC024909.1 | - | 8 | 995 | |||
BC056461.1 | - | 8 | 889 |