Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dera | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172733.1 | NP_766321.1 | 8 | 885 | ||
GenBank mRNA | AK012558.1 | - | 8 | 921 | ||
AK051983.1 | - | 1 | 1281 | |||
AK075963.1 | - | 8 | 910 | |||
AK083298.1 | - | 8 | 885 | |||
AK145820.1 | - | 8 | 921 | |||
AK150273.1 | - | 8 | 911 | |||
AK150712.1 | - | 8 | 881 | |||
AK152462.1 | - | 8 | 889 | |||
AK168204.1 | - | 8 | 921 | |||
AK169418.1 | - | 8 | 905 | |||
AK199271.1 | - | 3 | 252 | |||
BC016218.1 | - | 8 | 891 | 4484163 |