Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atp2c1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001253831.1 | NP_001240760.1 | 17 | 1635 | ||
NM_001253834.1 | NP_001240763.1 | 18 | 1726 | |||
NM_001253836.1 | NP_001240765.1 | 17 | 1658 | |||
NM_175025.4 | NP_778190.3 | 18 | 1841 | |||
GenBank mRNA | AJ551270.1 | - | 16 | 1423 | ||
AK028393.1 | - | 17 | 1604 | |||
AK133703.1 | - | 17 | 1636 | |||
AK159918.1 | - | 18 | 1792 | |||
AK161432.1 | - | 2 | 46 | |||
AK163049.1 | - | 18 | 1687 | |||
AK173154.1 | - | 16 | 1414 | |||
BC043091.1 | - | 18 | 1807 | 6405251 | ||
BC043706.1 | - | 18 | 1787 |