Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atp2c1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001253831.1 | NP_001240760.1 | 16 | 1538 | ||
NM_001253834.1 | NP_001240763.1 | 17 | 1629 | |||
NM_001253836.1 | NP_001240765.1 | 16 | 1561 | |||
NM_175025.4 | NP_778190.3 | 17 | 1744 | |||
GenBank mRNA | AJ551270.1 | - | 15 | 1326 | ||
AK028393.1 | - | 16 | 1507 | |||
AK133703.1 | - | 16 | 1539 | |||
AK159918.1 | - | 17 | 1695 | |||
AK163049.1 | - | 17 | 1590 | |||
AK173154.1 | - | 15 | 1317 | |||
BC043091.1 | - | 17 | 1710 | 6405251 | ||
BC043706.1 | - | 17 | 1690 |