Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ctdspl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290991.1 | NP_001277920.1 | 2 | 767 | ||
NM_001290992.1 | NP_001277921.1 | 2 | 767 | |||
NM_212450.4 | NP_997615.1 | 2 | 767 | |||
GenBank mRNA | AK030085.1 | - | 2 | 283 | ||
AK033382.1 | - | 2 | 316 | |||
AK041422.1 | - | 2 | 283 | |||
AK044805.1 | - | 2 | 296 | |||
AK049463.1 | - | 2 | 283 | |||
AK053513.1 | - | 2 | 296 | |||
AK082646.1 | - | 2 | 623 | |||
AK160431.1 | - | 2 | 282 | |||
AK163262.1 | - | 2 | 768 | |||
AK169248.1 | - | 2 | 309 | |||
BC052660.1 | - | 2 | 250 | 30049400 | ||
BC066219.1 | - | 2 | 288 |