Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kctd8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175519.5 | NP_780728.3 | 1 | 574 | ||
GenBank mRNA | AK039167.1 | - | 1 | 544 | ||
AK042569.1 | - | 1 | 427 | |||
AK043351.1 | - | 1 | 544 | |||
AK045439.1 | - | 1 | 427 | |||
AK047519.1 | - | 1 | 575 | |||
AK082563.1 | - | 1 | 428 | |||
AY615967.1 | - | 1 | 576 | |||
BC116940.1 | - | 1 | 225 | 40126259 | ||
BC116942.1 | - | 1 | 225 | 40126261 |