Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abcc12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172912.4 | NP_766500.3 | 9 | 1339 | ||
GenBank mRNA | AF502146.1 | - | 9 | 1339 | ||
AF514414.1 | - | 9 | 1339 | |||
AF514415.1 | - | 9 | 1339 | |||
AK030123.1 | - | 10 | 1460 | |||
AK133175.1 | - | 1429 | ||||
BC138380.1 | - | 9 | 1332 | 8861400 | ||
BC138381.1 | - | 9 | 1332 | 8861401 | ||
BC171952.1 | - | 9 | 1332 | 9054238 |