Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zswim8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252081.1 | NP_001239010.1 | 1 | 378 | ||
NM_001252082.1 | NP_001239011.1 | 1 | 378 | |||
NM_027996.3 | NP_082272.1 | 1 | 378 | |||
GenBank mRNA | AK053265.1 | - | 1 | 330 | ||
AK147398.1 | - | 1 | 379 | |||
BC151046.1 | - | 1 | 277 | 9087959 | ||
BC151056.1 | - | 1 | 277 | 9087969 | ||
BC151173.1 | - | 1 | 277 | 9088062 | ||
BC172103.1 | - | 1 | 277 | 9054389 |