Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lsamp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175548.3 | NP_780757.1 | 1 | 133 | ||
GenBank mRNA | AK030681.1 | - | 1 | 439 | ||
AK044845.1 | - | 1 | 133 | |||
AK140043.1 | - | 1 | 270 | |||
AK140663.1 | - | 1 | 132 | |||
AK158702.1 | - | 1 | 617 | |||
AK164171.1 | - | 440 | ||||
BC059067.1 | - | 1 | 649 |