Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: A230046K03Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033375.2 | NP_001028547.2 | 25, 26 | 2758 | ||
GenBank mRNA | AK038582.1 | - | 25, 26 | 2716 | ||
AK043911.1 | - | 25, 26 | 2729 | |||
AK129269.1 | - | 20, 21 | 2288 | |||
AK145000.1 | - | 25, 26 | 2740 | |||
AK161247.1 | - | 2, 3 | 206 | |||
AK166233.1 | - | 25, 26 | 2731 | |||
BC020314.2 | - | 22, 23 | 2393 | |||
BC026467.1 | - | 14, 15 | 1719 | |||
BC145215.1 | - | 25, 26 | 2953 |