Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PTPRD SIRNA Assays ›
Gene Symbol:
PTPRD
Gene Name:
protein tyrosine phosphatase, receptor type D
Gene Aliases:
HPTP, HPTPD, HPTPDELTA, PTPD, RPTPDELTA
Chromosome Location:
Chr.9: 8314246 - 10612840 on Build GRCh38
UniGene ID:
Hs.446083
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s11542
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PTPRD
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001040712.2
NP_001035802.1
9
1378
NM_001171025.1
NP_001164496.1
8
1357
NM_002839.3
NP_002830.1
21
2027
NM_130391.3
NP_569075.2
10
1387
NM_130392.3
NP_569076.2
10
1387
NM_130393.3
NP_569077.2
9
1369
XM_006716817.3
XP_006716880.1
20
2325
XM_006716823.2
XP_006716886.1
20
2319
XM_006716825.3
XP_006716888.1
20
2321
XM_006716827.3
XP_006716890.1
17
2279
XM_006716832.3
XP_006716895.1
18
2283
XM_006716833.3
XP_006716896.1
20
2315
XM_006716834.3
XP_006716897.1
17
2277
XM_006716835.3
XP_006716898.1
19
2306
XM_006716837.3
XP_006716900.1
19
2295
XM_006716838.3
XP_006716901.1
18
2288
XM_006716839.3
XP_006716902.1
17
2274
XM_011517992.2
XP_011516294.1
20
2318
XM_017014958.1
XP_016870447.1
20
2324
XM_017014959.1
XP_016870448.1
21
2396
XM_017014960.1
XP_016870449.1
19
2285
XM_017014961.1
XP_016870450.1
19
2269
XM_017014962.1
XP_016870451.1
11
1457
XM_017014963.1
XP_016870452.1
20
2324
XM_017014964.1
XP_016870453.1
20
2323
XM_017014965.1
XP_016870454.1
20
2326
XM_017014966.1
XP_016870455.1
19
2315
XM_017014967.1
XP_016870456.1
19
2310
XM_017014968.1
XP_016870457.1
20
2321
XM_017014969.1
XP_016870458.1
19
2306
XM_017014970.1
XP_016870459.1
20
2324
XM_017014971.1
XP_016870460.1
18
2295
XM_017014972.1
XP_016870461.1
20
2324
XM_017014973.1
XP_016870462.1
18
2291
XM_017014974.1
XP_016870463.1
20
2324
XM_017014975.1
XP_016870464.1
17
2280
XM_017014976.1
XP_016870465.1
20
2321
XM_017014977.1
XP_016870466.1
20
2322
XM_017014978.1
XP_016870467.1
20
2321
XM_017014979.1
XP_016870468.1
20
2323
XM_017014980.1
XP_016870469.1
20
2318
XM_017014981.1
XP_016870470.1
17
2274
XM_017014982.1
XP_016870471.1
20
2320
XM_017014983.1
XP_016870472.1
20
2320
XM_017014984.1
XP_016870473.1
19
2311
XM_017014985.1
XP_016870474.1
20
2317
XM_017014986.1
XP_016870475.1
20
2316
XM_017014987.1
XP_016870476.1
19
2298
XM_017014988.1
XP_016870477.1
19
2298
XM_017014989.1
XP_016870478.1
20
2316
XM_017014990.1
XP_016870479.1
20
2315
XM_017014991.1
XP_016870480.1
20
2315
XM_017014992.1
XP_016870481.1
19
2306
XM_017014993.1
XP_016870482.1
20
2313
XM_017014994.1
XP_016870483.1
20
2313
XM_017014995.1
XP_016870484.1
19
2310
GenBank mRNA
AB208869.1
-
8
1204
AB211400.1
-
20
1995
BC106713.1
-
8
1305
40027581
BC106714.1
-
10
1335
40027582
BC106715.2
-
10
1335
40027584
BC106716.1
-
9
1317
40027585
L38929.1
-
11
1437
X54133.1
-
1
117
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
protein dephosphorylation
phosphate-containing compound metabolic process
heterophilic cell-cell adhesion via plasma membrane cell adhesion molecules
transmembrane receptor protein tyrosine phosphatase signaling pathway
chemical synaptic transmission
neuron differentiation
peptidyl-tyrosine dephosphorylation
positive regulation of dendrite morphogenesis
presynaptic membrane assembly
transmembrane receptor protein tyrosine phosphatase activity
receptor binding
protein binding
cell adhesion molecule binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.31.0-2025.06.23-1.0