Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zscan10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033425.4 | NP_001028597.2 | 6 | 1000 | ||
NM_001289481.1 | NP_001276410.1 | 6 | 1000 | |||
NM_001289482.1 | NP_001276411.1 | 4 | 670 | |||
NM_001289483.1 | NP_001276412.1 | 6 | 496 | |||
NM_001289484.1 | NP_001276413.1 | 6 | 865 | |||
GenBank mRNA | AK141259.1 | - | 6 | 971 | ||
BC068284.1 | - | 5 | 673 | |||
BC145638.1 | - | 5 | 833 | 9054214 | ||
DQ323929.1 | - | 6 | 968 | |||
DQ323930.1 | - | 6 | 968 | |||
DQ323931.1 | - | 4 | 638 | |||
EF152498.1 | - | 5 | 815 | |||
EF152499.1 | - | 6 | 460 | |||
EF152501.1 | - | 4 | 592 | |||
EF152502.1 | - | 4 | 592 | |||
EF152503.1 | - | 2 | 469 | |||
EF152504.1 | - | 3 | 526 |