Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adam26a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010085.2 | NP_034215.2 | 2 | 1510 | ||
GenBank mRNA | AF167404.1 | - | 2 | 1469 | ||
AK029756.1 | - | 2 | 1511 |
Gene Symbol: Adam26b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009547.2 | NP_001009547.1 | 2 | 1500 | ||
GenBank mRNA | KF144352.1 | - | 1 | 22 | ||
KF144353.1 | - | 1 | 22 |