Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other GM5168 SIRNA Assays ›
          Gene Symbol:
          Gm5168 Gm10058 Gm10096 Gm10147 Gm10230 Gm10486 Gm10487 Gm10488 Gm14632 Gm14819 Gm2012 Gm4297 Gm4836 Gm5934 Gm5935 Gm6121 Slx
          Gene Name:
          predicted gene 5168
          predicted gene 10058
          predicted gene 10096
          predicted gene 10147
          predicted gene 10230
          predicted gene 10486
          predicted gene 10487
          predicted gene 10488
          predicted gene 14632
          predicted gene 14819
          predicted gene 2012
          predicted gene 4297
          predicted gene 4836
          predicted gene 5934
          predicted gene 5935
          predicted gene 6121
          Sycp3 like X-linked
          Gene Aliases:
          EG382275
          ENSMUSG00000059047
          ENSMUSG00000061227, Xmr-like
          ENSMUSG00000064334
          ENSMUSG00000068103
          ENSMUSG00000073247
          ENSMUSG00000073253
          ENSMUSG00000073257
          OTTMUSG00000017531
          OTTMUSG00000018086
          -
          -
          -
          EG546272
          EG546282
          -
          EG664958, Xmr
          Chromosome Location:
          Chr.X: 26028214 - 26051041 on Build GRCm38
          Chromosome Location:
          Chr.X: 27924021 - 27946885 on Build GRCm38
          Chromosome Location:
          Chr.X: 28931807 - 28954702 on Build GRCm38
          Chromosome Location:
          Chr.X: 28584995 - 28607874 on Build GRCm38
          Chromosome Location:
          Chr.X: 27472033 - 27494914 on Build GRCm38
          Chromosome Location:
          Chr.X: 35067384 - 35090316 on Build GRCm38
          Chromosome Location:
          Chr.X: 30820542 - 30843469 on Build GRCm38
          Chromosome Location:
          Chr.X: 29976314 - 29999162 on Build GRCm38
          Chromosome Location:
          Chr.X: 30329715 - 30352646 on Build GRCm38
          Chromosome Location:
          Chr.X: 35404215 - 35427094 on Build GRCm38
          Chromosome Location:
          Chr.X: 26200882 - 26231323 on Build GRCm38
          Chromosome Location:
          Chr.X: 24552250 - 24573305 on Build GRCm38
          Chromosome Location:
          Chr.X: 28270710 - 28293632 on Build GRCm38
          Chromosome Location:
          Chr.X: 24474308 - 24499163 on Build GRCm38
          Chromosome Location:
          Chr.X: 24753162 - 24775164 on Build GRCm38
          Chromosome Location:
          Chr.X: 26912743 - 26935669 on Build GRCm38
          Chromosome Location:
          Chr.X: 26522657 - 26545565 on Build GRCm38
          UniGene ID:
          Mm.370361
          Mm.483167
          -
          -
          Mm.483167
          Mm.458011
          Mm.427330
          Mm.432195
          Mm.483167
          Mm.483167
          Mm.484125
          -
          Mm.427330
          -
          Mm.391219
          Mm.483167
          Mm.489202
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          siRNA ID s118131

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Gm5168
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001025607.3 NP_001020778.1 4, 5 280
          GenBank mRNA BC099542.1 - 4, 5 280 30917040
          Gene Symbol: Gm10058
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001109969.2 NP_001103439.1 4, 5 330
          GenBank mRNA BU936093.1 - 333
          Gene Symbol: Gm10096
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001102678.2 NP_001096148.1 4, 5 361
          Gene Symbol: Gm10147
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001099919.2 NP_001093389.1 333
          Gene Symbol: Gm10230
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001099347.2 NP_001092817.1 4, 5 330
          Gene Symbol: Gm10486
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001109970.1 NP_001103440.1 4, 5 361
          GenBank mRNA BC087930.1 - 330 6773503
          BC099540.1 - 4, 5 301 6774523
          X72697.1 - 348
          Gene Symbol: Gm10487
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001100609.1 NP_001094079.1 4, 5 361
          Gene Symbol: Gm10488
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001099325.2 NP_001092795.1 4, 5 330
          GenBank mRNA BC099541.1 - 4, 5 330 30916030
          Gene Symbol: Gm14632
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001100610.2 NP_001094080.1 4, 5 330
          Gene Symbol: Gm14819
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001110250.2 NP_001103720.1 330
          Gene Symbol: Gm2012
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001104946.2 NP_001098416.1 4, 5 330
          Gene Symbol: Gm4297
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001100446.2 NP_001093916.1 3, 4 221
          Gene Symbol: Gm4836
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_009529.3 NP_033555.1 4, 5 361
          GenBank mRNA BC061169.1 - 4, 5 361 6774630
          Gene Symbol: Gm5934
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001100444.2 NP_001093914.1 3, 4 224
          Gene Symbol: Gm5935
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001081657.1 NP_001075126.1 4, 5 330
          GenBank mRNA BC100411.1 - 4, 5 330 30916114
          Gene Symbol: Gm6121
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001114754.1 NP_001108226.1 4, 5 330
          Gene Symbol: Slx
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001136476.1 NP_001129948.1 4, 5 361

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          spermatid development
          biological_process
          single fertilization
          sex determination
          regulation of gene expression
          spermatid differentiation
          molecular_function

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7b8d7ffddb-2cdg5:80/100.66.130.62:80.
          git-commit: 44eaffc35a31f29f64b20d0c2dd51bfa522c97f5
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0