Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Znrf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199143.2 | NP_954594.1 | 3, 4 | 756 | ||
GenBank mRNA | AF513708.1 | - | 3, 4 | 756 | ||
BC104316.2 | - | 3, 4 | 401 | |||
BC104317.2 | - | 3, 4 | 410 | |||
BC125242.1 | - | 3, 4 | 410 |