Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mettl21e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207281.3 | NP_997164.1 | 5 | 544 | ||
GenBank mRNA | AK029331.1 | - | 5 | 545 | ||
BC100531.1 | - | 5 | 487 | 30292619 |