Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scgb1b29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256066.1 | NP_001242995.1 | 2 | 146 | ||
GenBank mRNA | GU269228.1 | - | 2 | 146 |
Gene Symbol: Scgb1b19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281526.1 | NP_001268455.1 | 2 | 146 |
Gene Symbol: Scgb1b20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270543.1 | NP_001257472.1 | 2 | 162 | ||
GenBank mRNA | CK616096.1 | - | 2 | 163 | ||
CK616412.1 | - | 2 | 158 |
Gene Symbol: Scgb1b3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256073.1 | NP_001243002.1 | 2 | 162 | ||
GenBank mRNA | CK616146.1 | - | 2 | 180 |
Gene Symbol: Scgb1b7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270542.1 | NP_001257471.1 | 2 | 163 | ||
GenBank mRNA | CK615999.1 | - | 2 | 164 |