Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other RGS7 SIRNA Assays ›
          Gene Symbol:
          RGS7
          Gene Name:
          regulator of G-protein signaling 7
          Gene Aliases:
          -
          Chromosome Location:
          Chr.1: 240775193 - 241357374 on Build GRCh38
          UniGene ID:
          Hs.655739
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s11995

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: RGS7
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001282773.1 NP_001269702.1 12 1229
          NM_001282775.1 NP_001269704.1 14 1388
          NM_001282778.1 NP_001269707.1 14 1388
          NM_002924.5 NP_002915.3 14 1388
          XM_005273218.3 XP_005273275.1 14 1532
          XM_006711800.3 XP_006711863.1 13 1323
          XM_011544246.2 XP_011542548.1 13 982
          XM_011544247.2 XP_011542549.1 13 1027
          XM_017002001.1 XP_016857490.1 13 1481
          XM_017002002.1 XP_016857491.1 15 1407
          XM_017002003.1 XP_016857492.1 13 1481
          XM_017002004.1 XP_016857493.1 13 1481
          XM_017002005.1 XP_016857494.1 14 2423
          XM_017002006.1 XP_016857495.1 15 1453
          XM_017002007.1 XP_016857496.1 15 1384
          XM_017002008.1 XP_016857497.1 15 1412
          XM_017002009.1 XP_016857498.1 13 1312
          XM_017002010.1 XP_016857499.1 14 1097
          XM_017002011.1 XP_016857500.1 11 2494
          XM_017002012.1 XP_016857501.1 11 2492
          XM_017002013.1 XP_016857502.1 11 2491
          GenBank mRNA AF090116.1 - 14 1243
          AF090117.1 - 7 499
          AF493930.1 - 11 847
          AF493931.1 - 13 1006
          AK294234.1 - 14 1222
          AK294354.1 - 14 1363
          AK314862.1 - 14 1249
          AY587875.1 - 13 1006
          BC022009.1 - 14 1341 4823760
          U32439.1 - 12 860

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          protein folding
          G-protein coupled receptor signaling pathway
          negative regulation of signal transduction
          intracellular signal transduction
          positive regulation of GTPase activity
          signal transducer activity
          GTPase activator activity
          G-protein beta-subunit binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5bbdb6fccc-rsc2d:80/100.66.130.62:80.
          git-commit: 26de9a04ea7719c3b15362b12d28d672cac851f3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix