Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trim43a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034906.2 | NP_001030078.1 | 3 | 626 | ||
GenBank mRNA | AK145647.1 | - | 3 | 627 | ||
AK166678.1 | - | 3 | 625 | |||
BC141236.1 | - | 3 | 505 | 9055848 |
Gene Symbol: Trim43b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170884.1 | NP_001164355.1 | 3 | 771 |
Gene Symbol: Trim43c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177858.1 | NP_001171329.1 | 3 | 771 |