Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Triml2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001160412.1 | NP_001153884.1 | 7 | 835 | ||
GenBank mRNA | AV448075.1 | - | 5 | 338 | ||
AV467090.1 | - | 4 | 354 | |||
EU599038.1 | - | 9 | 925 | |||
EU599039.1 | - | 8 | 909 | |||
EU599040.1 | - | 7 | 768 |