Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CK137956 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134733.1 | NP_001128205.1 | 3 | 1244 | ||
GenBank mRNA | BC147529.1 | - | 3 | 1244 | 9087439 |