Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gcnt7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039560.3 | NP_001034649.1 | 2 | 833 | ||
GenBank mRNA | AK039414.1 | - | 2 | 835 | ||
AK132647.1 | - | 1 | 506 | |||
BC139073.1 | - | 2 | 793 | 8862095 | ||
BC139076.1 | - | 2 | 793 | 8862098 |