Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: I830012O16Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005858.3 | NP_001005858.2 | 967 | |||
GenBank mRNA | AK077243.1 | - | 956 | |||
AK150632.1 | - | 2 | 1214 | |||
AK151442.1 | - | 2 | 1214 | |||
AK152620.1 | - | 2 | 1214 |
Gene Symbol: Ifit3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010501.2 | NP_034631.1 | 2 | 1001 | ||
GenBank mRNA | AK077459.1 | - | 2 | 1000 | ||
AK151032.1 | - | 2 | 1001 | |||
AK153226.1 | - | 2 | 1002 | |||
BC003804.1 | - | 2 | 967 | 3487345 | ||
BC089563.1 | - | 2 | 999 | 30317428 | ||
CT010266.1 | - | 1 | 930 | |||
L32974.1 | - | 2 | 982 |