Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gm15319 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177408.1 | NP_001170879.1 | 1 | 325 | ||
GenBank mRNA | BC150794.1 | - | 1 | 177 | 9087705 |
Gene Symbol: 4930467E23Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039553.2 | NP_001034642.2 | 1 | 325 |
Gene Symbol: Gm21119 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270553.1 | NP_001257482.1 | 1 | 325 | ||
GenBank mRNA | AK076593.1 | - | 1 | 326 |